Proteintech
Manufacturer Code:138591AP150UL
Catalog # 50-172-9047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TIMM44 Fusion Protein Ag4834 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MIMT44 TIM44 TIMM44; Mitochondrial import inner membrane translocase subunit TIM44; translocase of inner mitochondrial membrane 44 homolog; translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed
Gene Aliases: 0710005E20Rik; D8Ertd118e; MIMT44; TIM44; TIMM44
UniProt ID: (Human) O43615, (Mouse) O35857, (Rat) O35094
Entrez Gene ID: (Human) 10469, (Mouse) 21856, (Rat) 29635
Molecular Function:
mitochondrial carrier protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.