Novus Biologicals
Manufacturer Code:NBP297571AF700
Catalog # NB045057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human TIMM17A fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inner membrane preprotein translocase Tim17a; Inner membrane preprotein translocase Tim17a mitochondrial import inner membrane translocase subunit Tim17-A mitochondrial inner membrane translocase TIM17Apreprotein translocase TIM17MIMT17 TIMM17 translocase of inner mitochondrial membrane 17 homolog A (yeast); Mitochondrial import inner membrane translocase subunit Tim17-A; mitochondrial inner membrane translocase; preprotein translocase
Gene Aliases: MIMT17; TIM17; TIM17A; TIMM17; TIMM17A
UniProt ID: (Human) Q99595
Entrez Gene ID: (Human) 10440
Molecular Function:
mitochondrial carrier protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.