Novus Biologicals
Manufacturer Code:NBP29402420UL
Catalog # NB022613
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-171 of human TIMM17A (NP_006326.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inner membrane preprotein translocase Tim17a; Inner membrane preprotein translocase Tim17a mitochondrial import inner membrane translocase subunit Tim17-A mitochondrial inner membrane translocase TIM17Apreprotein translocase TIM17MIMT17 TIMM17 translocase of inner mitochondrial membrane 17 homolog A (yeast); Mitochondrial import inner membrane translocase subunit Tim17-A; mitochondrial inner membrane translocase; preprotein translocase
Gene Aliases: MIMT17; TIM17; TIM17A; TIMM17; TIMM17A
UniProt ID: (Human) Q99595
Entrez Gene ID: (Human) 10440
Molecular Function:
mitochondrial carrier protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.