Invitrogen
Manufacturer Code:PA576173
Catalog # PIPA576173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human TIMM10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim10; small zinc finger-like protein; TIM10 TIM10A; translocase of inner mitochondrial membrane 10 homolog; translocase of inner mitochondrial membrane 13 homolog a
Gene Aliases: TIM10; TIM10A; Tim13; TIMM10; Timm13a
UniProt ID: (Human) P62072, (Rat) P62074, (Mouse) P62073
Entrez Gene ID: (Human) 26519, (Rat) 64464, (Mouse) 30059
Molecular Function: mitochondrial carrier protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.