Invitrogen
Manufacturer Code:PA592175
Catalog # PIPA592175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 969-1208 of human TIMELESS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTIM; mTim; Protein timeless homolog; rTIM; rTLP; timeless circadian clock 1; timeless homolog; timeless {ECO:0000312|RGD:620939}; Timeless-like protein; Tof1 homolog
Gene Aliases: C77407; Debt69; hTIM; TIM; TIM1; TIMELESS; TIMELESS1
UniProt ID: (Human) Q9UNS1, (Mouse) Q9R1X4, (Rat) Q9Z2Y1
Entrez Gene ID: (Human) 8914, (Mouse) 21853, (Rat) 83508
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.