Invitrogen
Manufacturer Code:PA578270
Catalog # PIPA578270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human TIMELESS. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTIM; mTim; Protein timeless homolog; TIM timeless circadian clock TIM1 hTIM; timeless circadian clock 1; timeless homolog; Tof1 homolog
Gene Aliases: C77407; Debt69; hTIM; TIM; TIM1; TIMELESS; TIMELESS1
UniProt ID: (Human) Q9UNS1, (Mouse) Q9R1X4
Entrez Gene ID: (Human) 8914, (Mouse) 21853
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.