Proteintech
Manufacturer Code:149271AP150UL
Catalog # 50-172-9842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TIM22 Fusion Protein Ag6689 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim22; preprotein translocase; putative membrane protein; Testis expressed sequence 4 TEX4 TIM22 TIMM22; Testis-expressed protein 4; testis-expressed sequence 4; translocase of inner mitochondrial membrane 22 homolog
Gene Aliases: TEX4; TIM22; TIMM22
UniProt ID: (Human) Q9Y584, (Mouse) Q9CQ85, (Rat) Q9JKW1
Entrez Gene ID: (Human) 29928, (Mouse) 56322, (Rat) 79463
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.