Invitrogen
Manufacturer Code:PA552690
Catalog # PIPA552690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TIM17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inner membrane preprotein translocase Tim17a; Mitochondrial import inner membrane translocase subunit Tim17-A; mitochondrial inner membrane translocase; preprotein translocase; TIM17 TIM17A inner membrane preprotein translocase Tim17a mitochondrial import inner membrane translocase subunit Tim17-A mitochondrial inner membrane translocase preprotein translocase
Gene Aliases: MIMT17; TIM17; TIM17A; TIMM17; TIMM17A
UniProt ID: (Human) Q99595
Entrez Gene ID: (Human) 10440
Molecular Function: mitochondrial carrier protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.