R&D Systems
Manufacturer Code:MAB23652100
Catalog # MAB23652100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2321C |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TIM-3 Met1-Arg200 Accession No. Q8TDQ0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
Protein Aliases: CD366; CD366 FLJ14428 HAVcr-2 HAVCR2 hepatitis A virus cellular receptor 2 kidney injury molecule-3 KIM-3 SPTCL T cell immunoglobulin mucin 3 T cell immunoglobulin mucin-3 T-cell immunoglobulin and mucin domain-containing protein 3 TIM 3 Tim-3 TIM3 TIM3 T-cell membrane protein 3 TIMD3 TIMD-3 TIMD3KIM-3; HAVcr-2; Hepatitis A virus cellular receptor 2; kidney injury molecule-3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; TIM-3; TIMD-3
Gene Aliases: CD366; HAVcr-2; HAVCR2; KIM-3; Tim-3; TIM3; TIMD-3; TIMD3
UniProt ID: (Human) Q8TDQ0
Entrez Gene ID: (Human) 84868
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.