Novus Biologicals
Manufacturer Code:NBP149534AF350
Catalog # NP14934A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88] |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: chromosome 12 open reading frame 5 EC 3.1.3.46 probable fructose-26-bisphosphatase TIGAR TIGARTP53-induced glycolysis and apoptosis regulator transactivated by NS3TP2 protein; Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; probable fructose-2,6-bisphosphatase TIGAR; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein
Gene Aliases: C12orf5; FR2BP; TIGAR
UniProt ID: (Human) Q9NQ88
Entrez Gene ID: (Human) 57103
Molecular Function:
isomerase
mutase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.