Invitrogen
Manufacturer Code:PA520347
Catalog # PIPA520347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide from near the center of human TIGAR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; probable fructose-2,6-bisphosphatase TIGAR; TIGAR TP53-induced glycolysis and apoptosis regulator probable fructose-26-bisphosphatase TIGAR transactivated by NS3TP2 protein; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein
Gene Aliases: 9630033F20Rik; AA793651; AI595337; C12orf5; C79710; C85509; FR2BP; TIGAR
UniProt ID: (Human) Q9NQ88, (Mouse) Q8BZA9
Entrez Gene ID: (Human) 57103, (Mouse) 319801
Molecular Function:
isomerase
mutase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.