Novus Biologicals
Manufacturer Code:NBP274514JF646
Catalog # N274514J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D9 |
Immunogen | Recombinant protein fragment corresponding to amino acids 706-1016 of human TIF1 alpha (NP_003843) produced in E.coli. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: E3 ubiquitin-protein ligase TRIM24; E3 ubiquitin-protein ligase TRIM24 EC 6.3.2 EC 6.3.2.- hTIF1 PTC6 RING finger protein 82 RNF82Tif1a TIF1-alpha TIF1ATIF1ALPHA TIF1TF1A transcription intermediary factor 1-alpha transcriptional intermediary factor 1 tripartite motif containing 24 tripartite motif-containing 24 Tripartite motif-containing protein 24; RING finger protein 82; RING-type E3 ubiquitin transferase TIF1-alpha; TIF1-alpha; Transcription intermediary factor 1-alpha; transcriptional intermediary factor 1; tripartite motif-containing 24; Tripartite motif-containing protein 24
Gene Aliases: hTIF1; PTC6; RNF82; TF1A; TIF1; TIF1A; TIF1ALPHA; TRIM24
UniProt ID: (Human) O15164
Entrez Gene ID: (Human) 8805
Molecular Function:
transcription cofactor
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.