Proteintech
Manufacturer Code:232881AP150UL
Catalog # 50-173-4038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIF/TICAM1 Fusion Protein Ag19779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB TICAM 1 TICAM1 TRIF; Putative NF-kappa-B-activating protein 502H; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1
Gene Aliases: IIAE6; MyD88-3; PRVTIRB; TICAM-1; TICAM1; TRIF
UniProt ID: (Human) Q8IUC6
Entrez Gene ID: (Human) 148022
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.