Novus Biologicals
Manufacturer Code:NBP274513AF405
Catalog # N274513A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D7 |
Immunogen | Full length recombinant protein of human TIA1 (NP_071505) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cytotoxic granule-associated RNA-binding protein nucleolysin TIA-1 isoform p40 p40-TIA-1 p40-TIA-1 (containing p15-TIA-1) RNA-binding protein TIA-1 T-cell-restricted intracellular antigen-1 TIA-1 TIA1 cytotoxic granule-associated RNA binding protein TIA1 cytotoxic granule-associated RNA-binding protein; Nucleolysin TIA-1 isoform p40; p40-TIA-1; p40-TIA-1 (containing p15-TIA-1); RNA-binding protein TIA-1; T-cell-restricted intracellular antigen-1; TIA-1
Gene Aliases: TIA-1; TIA1; WDM
UniProt ID: (Human) P31483
Entrez Gene ID: (Human) 7072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.