Novus Biologicals
Manufacturer Code:NBP274511FR
Catalog # NBP274511FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI13E3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 2-230 of human THTPA (NP_077304) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.6.1.28 MGC2652 thiamine triphosphatase thiamine-triphosphatase THTP THTPASE; Thiamine-triphosphatase; ThTPase
Gene Aliases: THTP; THTPA; THTPASE
UniProt ID: (Human) Q9BU02
Entrez Gene ID: (Human) 79178
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.