Abnova
Manufacturer Code:MAB1784
Catalog # 89-110-626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2386 |
Immunogen | A synthetic peptide corresponding to N-terminus human THRB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of THRB.
For Research Use Only
Protein Aliases: c-erbA-2; c-erbA-beta; ERBA-BETA; ERBA2; GRTH; MGC126109; MGC126110; NR1A2; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; PRTH; THR1; THRB1; THRB2; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian)
Gene Aliases: C-ERBA-2; C-ERBA-BETA; ERBA2; GRTH; NR1A2; PRTH; THR1; THRB; THRB1; THRB2
UniProt ID: (Human) P10828
Entrez Gene ID: (Human) 7068
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.