Invitrogen
Manufacturer Code:PA568790
Catalog # PIPA568790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of mouse THRB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-erbA-2; c-erbA-beta; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; THRB thyroid hormone receptor beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2 avian) ERBA-BETA ERBA2 GRTH MGC126109 MGC126110 NR1A2 THR1 THRB1 THRB2 avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2 generalized resistance to thyroid hormone oncogene ERBA2 thyroid hormone receptor beta 1 thyroid hormone receptor beta; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor beta 3; thyroid hormone receptor beta2delta; thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2); thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); TR beta; TRbeta2Delta
Gene Aliases: C-ERBA-2; C-ERBA-BETA; c-erbAbeta; ERBA2; GRTH; NR1A2; PRTH; RATT3REC; T3R[b]; T3Rbeta; T3rec; THR1; THRB; THRB1; THRB2; TRbeta
UniProt ID: (Human) P10828, (Mouse) P37242
Entrez Gene ID: (Human) 7068, (Horse) 100051615, (Guinea pig) 100721125, (Mouse) 21834, (Rat) 24831
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.