Invitrogen
Manufacturer Code:PA138763
Catalog # 01-674-734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from N-terminus of human THR-beta protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-erbA-2; C-ERBA-2 C-ERBA-BETA ERBA2 GRTH NR1A2 PRTH THR1 THRB1 THRB2 nuclear receptor subfamily 1 group A member 2 oncogene ERBA2 thyroid hormone nuclear receptor beta variant 1 thyroid hormone receptor beta thyroid hormone receptor beta erythroblastic leukemia viral v-erb-a oncogene homolog 2 avian; c-erbA-beta; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor beta; thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian)
Gene Aliases: C-ERBA-2; C-ERBA-BETA; ERBA2; GRTH; NR1A2; PRTH; THR1; THRB; THRB1; THRB2
UniProt ID: (Human) P10828
Entrez Gene ID: (Human) 7068
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.