OriGene
Manufacturer Code:TA805258
Catalog # 50-168-0611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-220 of human THRA produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AR7 CHNG6 EAR7 ERB-T-1 ERBA ERBA1 NR1A1 THRA1 THRA2 c-ERBA-1; c-erbA-1; c-erbA-alpha; EAR-7; ERBA-related 7; Nuclear receptor subfamily 1 group A member 1; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; triiodothyronine receptor; V-erbA-related protein 7
Gene Aliases: AR7; c-ERBA-1; CHNG6; EAR7; ERB-T-1; ERBA; ERBA1; NR1A1; THRA; THRA1; THRA2
UniProt ID: (Human) A8K3B5
Entrez Gene ID: (Human) 7067
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.