Proteintech
Manufacturer Code:101391AP150UL
Catalog # 50-172-6054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | THRA Fusion Protein Ag0182 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AR7 c ERBA 1 c erbA alpha EAR 7 EAR7 ERB T 1 ERBA ERBA1 NR1A1 THRA THRA1 THRA2 Thyroid hormone receptor alpha V erbA related protein 7; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; c-erb-A thyroid hormone receptor; c-erbA-1; C-erbA-alpha; c-erbA-alpha2; c-erbAalpha; c-erbAalpha2; EAR-7; ERBA-related 7; Nuclear receptor subfamily 1 group A member 1; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid normone nuclear receptor alpha variant 1; TR alpha 1; TR alpha 2; triiodothyronine receptor; V-erbA-related protein 7
Gene Aliases: 6430529J03Rik; AR7; AW259572; c-ERBA-1; C-erba-alpha; CHNG6; EAR7; ERB-T-1; ERBA; ERBA1; NR1A1; Rvr; T3R[a]; T3Ralpha; THRA; THRA1; THRA2
UniProt ID: (Human) A8K3B5, (Mouse) P63058, (Rat) P63059
Entrez Gene ID: (Human) 7067, (Mouse) 21833, (Rat) 81812
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.