Proteintech
Manufacturer Code:109201AP150UL
Catalog # 50-172-6714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | THOC1 Fusion Protein Ag1352 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPR1 hTREX84 Nuclear matrix protein p84 P84 P84N5 THO complex 1 THO complex subunit 1 Tho1 THOC1; hTREX84; Nuclear matrix protein p84; p84N5; THO complex subunit 1; Tho1
Gene Aliases: 3110002N20Rik; AW107452; HPR1; LRRGT00070; NMP-84; P84; P84N5; THOC1
UniProt ID: (Human) Q96FV9, (Mouse) Q8R3N6
Entrez Gene ID: (Human) 9984, (Mouse) 225160, (Rat) 291797
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.