Abnova
Manufacturer Code:PAB27635
Catalog # 89-129-147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 192-204 at internal region of human THNSL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of THNSL2.
For Research Use Only
Protein Aliases: FLJ10916; secreted osteoclastogenic factor of activated T cells; Secreted osteoclastogenic factor of activated T-cells; SOFAT; Threonine synthase-like 2; THS2; TSH2
Gene Aliases: SOFAT; THNSL2; THS2; TSH2
UniProt ID: (Human) Q86YJ6
Entrez Gene ID: (Human) 55258
Molecular Function: deaminase dehydratase epimerase/racemase hydrolase isomerase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.