Proteintech
Manufacturer Code:175731AP150UL
Catalog # 50-173-1656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | THNSL2 Fusion Protein Ag11788 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: secreted osteoclastogenic factor of activated T cells; Secreted osteoclastogenic factor of activated T-cells; SOFAT; THNSL2 Threonine synthase like 2 THS2 TSH2; Threonine synthase-like 2; TSH2
Gene Aliases: BC051244; RGD1309144; SOFAT; THNSL2; THS2; TSH2
UniProt ID: (Human) Q86YJ6, (Mouse) Q80W22, (Rat) Q5M7T9
Entrez Gene ID: (Human) 55258, (Mouse) 232078, (Rat) 297332
Molecular Function: deaminase dehydratase epimerase/racemase hydrolase isomerase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.