Invitrogen
Manufacturer Code:MA524482
Catalog # PIMA524482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4F11 |
Immunogen | THG1L (AAH01523 1 a.a. &bsim 269 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IHG-1; induced by high glucose-1; Induced in high glucose-1; Interphase cytoplasmic foci protein 45; Probable tRNA(His) guanylyltransferase; THG1L ICF45; tRNA-histidine guanylyltransferase; tRNA-histidine guanylyltransferase 1-like
Gene Aliases: hTHG1; ICF45; IHG-1; THG1L
UniProt ID: (Human) Q9NWX6
Entrez Gene ID: (Human) 54974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.