Invitrogen
Manufacturer Code:PA517637
Catalog # PIPA517637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues of human CTMP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA thioesterase THEM4; Acyl-coenzyme A thioesterase THEM4; C-terminal modulator protein; C-terminal modulator protein carboxyl-terminal modulator protein; Carboxyl-terminal modulator protein; Thioesterase superfamily member 4
Gene Aliases: 2700077M13Rik; 4921507I02Rik; CTMP; RGD1304723; THEM4
UniProt ID: (Human) Q5T1C6, (Rat) Q566R0, (Mouse) Q3UUI3
Entrez Gene ID: (Human) 117145, (Rat) 361992, (Mouse) 75778
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.