Invitrogen
Manufacturer Code:PA5110484
Catalog # PIPA5110484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 37-150 of human THEM4 (NP_444283.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700077M13Rik 4921507I02Rik Acyl-CoA thioesterase THEM4 Acyl-coenzyme A thioesterase THEM4 Carboxyl-terminal modulator protein Ctmp RGD1304723 THEM4 Thioesterase superfamily member 4; Acyl-CoA thioesterase THEM4; Acyl-coenzyme A thioesterase THEM4; C-terminal modulator protein; Carboxyl-terminal modulator protein; Thioesterase superfamily member 4
Gene Aliases: 2700077M13Rik; 4921507I02Rik; CTMP; THEM4
UniProt ID: (Human) Q5T1C6, (Mouse) Q3UUI3
Entrez Gene ID: (Human) 117145, (Mouse) 75778
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.