Proteintech
Manufacturer Code:167821AP150UL
Catalog # 50-173-1175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | THEM2 Fusion Protein Ag10212 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACOT13 Acyl CoA thioesterase 13 HT012 PNAS 27 THEM2; Acyl-CoA thioesterase 13; Acyl-coenzyme A thioesterase 13; Acyl-coenzyme A thioesterase 13, N-terminally processed; Hotdog-fold thioesterase superfamily member 2; hypothalamus protein HT012; Palmitoyl-CoA hydrolase; THEM2; Thioesterase superfamily member 2
Gene Aliases: 0610006O17Rik; ACOT13; HT012; PNAS-27; THEM2
UniProt ID: (Human) Q9NPJ3, (Mouse) Q9CQR4
Entrez Gene ID: (Human) 55856, (Rat) 291135, (Mouse) 66834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.