R&D Systems
Manufacturer Code:MAB4286
Catalog # MAB4286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 409522 |
Immunogen | NS0 mouse myeloma cell line transfected with human TGR5/GPBAR1 Met1-Asn330 Accession # Q8TDU6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BG37; BG37 BG37hBG37 G protein-coupled bile acid receptor 1 GPBA GPBAR1 GPCR19 GPR131 G-protein coupled bile acid receptor 1 G-protein coupled bile acid receptor BG37 G-protein coupled receptor GPCR19 M-BAR M-BARhGPCR19 membrane bile acid receptor Membrane-type receptor for bile acids MGC40597 TGR5GPCR; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; hGPCR19; M-BAR; membrane bile acid receptor; Membrane-type receptor for bile acids
Gene Aliases: BG37; GPBAR1; GPCR19; GPR131; M-BAR; TGR5
UniProt ID: (Human) Q8TDU6
Entrez Gene ID: (Human) 151306
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.