Novus Biologicals
Manufacturer Code:NBP220614
Catalog # NBP220614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TGR5. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: BG37; BG37hBG37 G protein-coupled bile acid receptor 1 GPCR19 GPR131 G-protein coupled bile acid receptor 1 G-protein coupled bile acid receptor BG37 G-protein coupled receptor GPCR19 M-BARhGPCR19 membrane bile acid receptor Membrane-type receptor for bile acids MGC40597 TGR5GPCR; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; hGPCR19; M-BAR; membrane bile acid receptor; Membrane-type receptor for bile acids
Gene Aliases: BG37; GPBAR1; GPCR19; GPR131; M-BAR; TGR5
UniProt ID: (Human) Q8TDU6
Entrez Gene ID: (Human) 151306
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.