Invitrogen
Manufacturer Code:PA523183
Catalog # PIPA523183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (41 WDRRLRSPPA 50) of BG37/TGR5 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BG37; BG37 GPCR19 GPR131 M-BAR TGR5 G-protein coupled bile acid receptor 1 G-protein coupled bile acid receptor BG37 G-protein coupled receptor GPCR19 hBG37 hGPCR19 membrane bile acid receptor membrane-type receptor for bile acids; G protein-coupled receptor; G protein-coupled receptor TGR5; G-protein coupled bile acid receptor 1; G-protein coupled bile acid receptor BG37; G-protein coupled receptor GPCR19; hBG37; hGPCR19; M-BAR; membrane bile acid receptor; Membrane-type receptor for bile acids
Gene Aliases: BG37; GPBAR1; GPCR; GPCR19; GPR131; M-BAR; TGR5
UniProt ID: (Human) Q8TDU6, (Rat) Q80T02, (Mouse) Q80SS6
Entrez Gene ID: (Human) 151306, (Rat) 338443, (Mouse) 227289
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.