Proteintech
Manufacturer Code:250711AP150UL
Catalog # 50-173-4708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGM5 Fusion Protein Ag18492 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein-glutamine gamma-glutamyltransferase 5; TG(X); TG(X) TGase 5 TGase X TGM5 TGM6 TGMX TGX transglutaminase 5 Transglutaminase X; TGase 5; TGase X; TGase-5; transglutaminase V; Transglutaminase X; Transglutaminase-5
Gene Aliases: 2310007C07Rik; PSS2; TGASE5; TGASEX; TGM5; TGM6; TGMX; TGX
UniProt ID: (Human) O43548, (Mouse) Q9D7I9
Entrez Gene ID: (Human) 9333, (Mouse) 74176
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.