Proteintech
Manufacturer Code:158471AP150UL
Catalog # 50-173-0525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGM4 Fusion Protein Ag8608 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Experimental autoimmune prostatitis antigen 1; Fibrinoligase; Fibrinoligase hTGP Prostate transglutaminase TG(P) TGase 4 TGase P TGM4 TGP Transglutaminase 4 transglutaminase 4 (prostate) Transglutaminase P; Prostate transglutaminase; Prostate-specific transglutaminase; Protein-glutamine gamma-glutamyltransferase 4; TG(P); TGase P; TGase-4; tgm4; transglutaminase 4 (prostate); Transglutaminase P; Transglutaminase-4
Gene Aliases: 9530008N10Rik; Eapa1; hTGP; TGM4; TGP
UniProt ID: (Human) P49221, (Mouse) Q8BZH1
Entrez Gene ID: (Human) 7047, (Mouse) 331046
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.