Proteintech
Manufacturer Code:151001AP150UL
Catalog # 50-172-9978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGM2 Fusion Protein Ag7439 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C polypeptide; C polypeptide, protein-glutamine-gamma-glutamyltransferase; Erythrocyte transglutaminase; G ALPHA h GNAH TG(C) TG2 TGase 2 TGase C TGase H TGC TGM2 Tissue transglutaminase Transglutaminase 2 Transglutaminase C Transglutaminase H; G(h); Heart G alpha(h); hhG alpha(h); Isopeptidase TGM2; Protein G alpha(h); Protein G(h); Protein-glutamine deamidase TGM2; Protein-glutamine dopaminyltransferase TGM2; Protein-glutamine gamma-glutamyltransferase 2; Protein-glutamine histaminyltransferase TGM2; Protein-glutamine noradrenalinyltransferase TGM2; Protein-glutamine serotonyltransferase TGM2; protein-glutamine-gamma-glutamyltransferase; TG C; TG(C); TG2; TGase 2; TGase C; TGase H; TGase II; TGase-2; TGC; Tissue transglutaminase; tissue-type transglutaminase; transglutaminase 2, C polypeptide; Transglutaminase C; Transglutaminase H; Transglutaminase II; Transglutaminase-2; tTG
Gene Aliases: G[a]h; TG(C); TG2; TGase2; TgaseII; TGC; TGM2; tTG; tTGas
UniProt ID: (Human) P21980, (Rat) Q9WVJ6, (Mouse) P21981
Entrez Gene ID: (Human) 7052, (Rat) 56083, (Mouse) 21817
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.