Proteintech
Manufacturer Code:201531AP150UL
Catalog # 50-173-2736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGFBRAP1 Fusion Protein Ag13698 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TGF-beta receptor-associated protein 1; TGFBRAP1 TRAP 1 TRAP1; Transforming growth factor-beta receptor-associated protein 1; VPS3 CORVET complex subunit
Gene Aliases: TGFBRAP1; TRAP-1; TRAP1; VPS3
UniProt ID: (Human) Q8WUH2
Entrez Gene ID: (Human) 9392
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.