Invitrogen
Manufacturer Code:PA5116035
Catalog # PIPA5116035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TGF beta Receptor III corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110036H20Rik AU015626 AW215636 betaglycan betaglycan proteoglycan BGCAN OTTHUMP00000011787 TBRIII tgf beta receptor 3 TGF-beta receptor type 3 TGF-beta receptor type III Tgfbr3 TGFR-3 transforming growth factor beta receptor 3 transforming growth factor beta receptor III Transforming growth factor beta receptor type 3 transforming growth factor beta receptor 3 transforming growth factor beta receptor III; Betaglycan; betaglycan proteoglycan; TGF-beta receptor type 3; TGF-beta receptor type III; TGFR-3; Transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; transforming growth factor, beta receptor 3
Gene Aliases: 1110036H20Rik; AU015626; AW215636; betaglycan; BGCAN; TBRIII; TGFBR3
UniProt ID: (Human) Q03167, (Mouse) O88393, (Rat) P26342
Entrez Gene ID: (Human) 7049, (Mouse) 21814, (Rat) 29610
Molecular Function:
TGF-beta receptor
cytokine receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.