Abnova
Manufacturer Code:PAB17995
Catalog # 89-119-596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the residues surrounding amino acid 555 of rat Tgfbr2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Tgfbr2.
For Research Use Only
Protein Aliases: tbetaR-II; TGF-beta 2; TGF-beta receptor type II; TGF-beta receptor type-2; TGF-beta type II receptor; Tgfbr2T; TGFR-2; transforming growth factor beta receptor type II; transforming growth factor, beta receptor II; transforming growth factor, beta receptor IIT; transforming growth factor-b type II receptor; Transforming growth factor-beta receptor type II; transforming growth factor-beta type II receptor
Gene Aliases: TGF-beta 2; Tgfbr2; Tgfbr2T
UniProt ID: (Rat) P38438
Entrez Gene ID: (Rat) 81810
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.