Invitrogen
Manufacturer Code:PA538718
Catalog # PIPA538718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic phosphopeptide derived from human TGF beta Receptor I around the phosphorylation site of Ser165 (D-P-SP-L-D) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAT5 ACVRLK4 ALK-5 ALK5 LDS1A LDS2A MSSE SKR4 TGFR-1 TGF-beta receptor type I TGF-beta receptor type-1 TGF-beta type I receptor activin A receptor type II-like kinase 53kD activin A receptor type II-like kinase 53kDa activin A receptor type II-like protein kinase of 53kD TGF B Receptor 1 TGF B Receptor I; activin A receptor type II-like kinase, 53kDa; Activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ALK-5; ESK2; mutant transforming growth factor beta receptor I; Serine/threonine-protein kinase receptor R4; SKR4; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFR-1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; transforming growth factor, beta receptor I; Transforming growth factor-beta receptor type I
Gene Aliases: AAT5; ACVRLK4; ALK-5; ALK5; AU017191; ESS1; LDS1; LDS1A; LDS2A; MSSE; SKR4; TbetaR-I; TbetaRI; TGFBR1; TGFR-1
UniProt ID: (Human) P36897, (Rat) P80204, (Mouse) Q64729
Entrez Gene ID: (Human) 7046, (Rat) 29591, (Mouse) 21812
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.