Abnova
Manufacturer Code:MAB12310
Catalog # 89-935-001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 220ct16.4.3.1 |
Immunogen | Recombinant protein corresponding to human TGFB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against human TGFB2.
For Research Use Only
Protein Aliases: BSC-1 cell growth inhibitor; Cetermin; G-TSF; Glioblastoma-derived T-cell suppressor factor; LAP; Latency-associated peptide; MGC116892; polyergin; prepro-transforming growth factor beta-2; TGF-beta-2; TGF-beta2; Transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor, beta 2
Gene Aliases: LDS4; TGF-beta2; TGFB2
UniProt ID: (Human) P61812
Entrez Gene ID: (Human) 7042
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.