Novus Biologicals
Manufacturer Code:NBP290633AF488
Catalog # NB075844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 30 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse TGF-beta RIII (NP_072075.2 Met1-Arg399). |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Betaglycan; betaglycan betaglycan proteoglycan BGCAN TGF-beta receptor type 3 TGF-beta receptor type III TGFR-3 Transforming growth factor beta receptor III transforming growth factor beta receptor type 3 transforming growth factor beta receptor III transforming growth factor beta receptor III (betaglycan 300kDa); betaglycan proteoglycan; TGF-beta receptor type 3; TGF-beta receptor type III; TGFR-3; Transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3
Gene Aliases: betaglycan; BGCAN; TGFBR3
UniProt ID: (Human) Q03167
Entrez Gene ID: (Human) 7049
Molecular Function: TGF-beta receptor cytokine receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.