R&D Systems
Manufacturer Code:FAB532C025
Catalog # FAB532C025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse TGF-beta RII and <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant mouse TGF-beta RII Ile24-Asp184 Accession # Q62312 |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
For Research Use Only
Protein Aliases: AAT3 EC 2.7.11 EC 2.7.11.30 FAA3 HNPCC6 LDS1B LDS2B MFS2 RIIC TAAD2 tbetaR-II TGF-beta receptor type II TGF-beta receptor type IIB TGF-beta receptor type-2 TGF-beta type II receptor TGFbetaRII TGFbeta-RII TGFBR2 TGF-bRII TGFR-2 transforming growth factor beta receptor type IIC transforming growth factor beta receptor II (70/80kDa) transforming growth factor beta receptor II (70/80kDa) isoform 1 transforming growth factor beta receptor II (70/80kDa) isoform 2 transforming growth factor beta receptor II (70-80kD) Transforming growth factor-beta receptor type II; tbetaR-II; TGF-beta receptor type II; TGF-beta receptor type IIB; TGF-beta receptor type-2; TGF-beta type II receptor; TGFR-2; transforming growth factor beta receptor II; transforming growth factor beta receptor type IIC; transforming growth factor, beta receptor II (70/80kDa); transforming growth factor, beta receptor II alpha; transforming growth factor, beta receptor II beta; transforming growth factor, beta receptor II delta; transforming growth factor, beta receptor II epsilon; transforming growth factor, beta receptor II gamma; Transforming growth factor-beta receptor type II
Gene Aliases: AAT3; FAA3; LDS1B; LDS2; LDS2B; MFS2; RIIC; TAAD2; TGFbeta-RII; TGFBR2; TGFR-2
UniProt ID: (Human) P37173
Entrez Gene ID: (Human) 7048
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.