Novus Biologicals
Manufacturer Code:NBP211921AF647
Catalog # NB211921647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0016-3A11 |
Immunogen | Mouse TGF beta Receptor I |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: AAT5 activin A receptor type II-like kinase 53kD activin A receptor type II-like kinase 53kDa Activin receptor-like kinase 5 ACVRLK4 ALK-5ALK5 EC 2.7.11 EC 2.7.11.30 LDS1A LDS2A Serine/threonine-protein kinase receptor R4 SKR4 tbetaR-I TGFB1R1 TGF-beta receptor type I TGF-beta receptor type-1 TGF-beta type I receptor TGFR-1 transforming growth factor beta receptor I transforming growth factor beta receptor 1 transforming growth factor beta receptor I (activin A receptor type II-likekinase 53kD) Transforming growth factor-beta receptor type I; activin A receptor type II-like kinase, 53kDa; Activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ALK-5; mutant transforming growth factor beta receptor I; Serine/threonine-protein kinase receptor R4; SKR4; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFR-1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; Transforming growth factor-beta receptor type I
Gene Aliases: AAT5; ACVRLK4; ALK-5; ALK5; ESS1; LDS1; LDS1A; LDS2A; MSSE; SKR4; tbetaR-I; TGFBR1; TGFR-1
UniProt ID: (Human) P36897
Entrez Gene ID: (Human) 7046
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.