R&D Systems
Manufacturer Code:AF3025
Catalog # AF3025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TGF-beta RI/ALK-5 Leu34-Glu125 Accession # P36897 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AAT5 activin A receptor type II-like kinase 53kD activin A receptor type II-like kinase 53kDa Activin receptor-like kinase 5 ACVRLK4 ALK-5 ALK-5ALK5 EC 2.7.11 EC 2.7.11.30 LDS1A LDS2A Serine/threonine-protein kinase receptor R4 SKR4 TbetaR-I TGFB1R1 TGF-beta receptor type I TGF-beta receptor type-1 TGF-beta type I receptor TGFbetaRI TGFBR1 TGF-bRI TGFR-1 transforming growth factor beta receptor I transforming growth factor beta receptor 1 transforming growth factor beta receptor I (activin A receptor type II-likekinase 53kD) Transforming growth factor-beta receptor type I; activin A receptor type II-like kinase, 53kDa; Activin A receptor type II-like protein kinase of 53kD; Activin receptor-like kinase 5; ALK-5; mutant transforming growth factor beta receptor I; Serine/threonine-protein kinase receptor R4; SKR4; TGF-beta receptor type I; TGF-beta receptor type-1; TGF-beta type I receptor; TGFR-1; transforming growth factor beta receptor I; transforming growth factor, beta receptor 1; Transforming growth factor-beta receptor type I
Gene Aliases: AAT5; ACVRLK4; ALK-5; ALK5; ESS1; LDS1; LDS1A; LDS2A; MSSE; SKR4; tbetaR-I; TGFBR1; TGFR-1
UniProt ID: (Human) P36897
Entrez Gene ID: (Human) 7046
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.