R&D Systems
Manufacturer Code:BAM243
Catalog # BAM243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 20724 |
Immunogen | recombinant human (rh) TGF-beta 3 Ala301-Ser412 (Tyr340Phe) accession # P10600 mixed with rhTGF-beta 1 rhTGF-beta 2 recombinant canine TGF-beta 3 and recombinant amphibian TGF-beta 5 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARVD ARVD1 FLJ16571 LDS5 RNHF TGFB3 TGFbeta 3 TGF-beta3 TGF-beta-3 transforming growth factor beta-3 transforming growth factor beta 3; LAP; Latency-associated peptide; prepro-transforming growth factor beta-3; TGF-beta-3; Transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3
Gene Aliases: ARVD; ARVD1; LDS5; RNHF; TGF-beta3; TGFB3
UniProt ID: (Human) P10600
Entrez Gene ID: (Human) 7043
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.