Invitrogen
Manufacturer Code:PA578197
Catalog # PIPA578197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human TGF beta 3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAP; Latency-associated peptide; prepro-transforming growth factor beta-3; TGF-beta-3; TGFB3 ARVD transforming growth factor beta 3 TGF-beta3; Transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3
Gene Aliases: ARVD; ARVD1; LDS5; RNHF; TGF-beta3; Tgfb-3; TGFB3
UniProt ID: (Human) P10600
Entrez Gene ID: (Human) 7043, (Mouse) 21809
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.