Invitrogen
Manufacturer Code:PA112642
Catalog # 01-674-805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to the extracellular domain of the chicken TGF III receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: LAP; Latency-associated peptide; TGF beta 3; TGF-beta-3; TGF-beta3; transforming growth factor beta precursor; Transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor-beta3
Gene Aliases: TGF-beta-3; TGF-beta3; TGFB3
UniProt ID: (Chicken) P16047
Entrez Gene ID: (Chicken) 396438
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.