Invitrogen
Manufacturer Code:PA5112839
Catalog # PIPA5112839
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Transforming growth factor beta-2 proprotein protein (304-413AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BB105277 BSC-1 cell growth inhibitor Cetermin Glioblastoma-derived T-cell suppressor factor G-TSF LAP Latency-associated peptide LDS4 polyergin prepro-transforming growth factor beta-2 TGF beta 2 protein TGFB TGFB2 TGF-B2 Tgfb-2 Tgf-beta2 TGF-beta-2 Transforming growth factor transforming growth factor beta 2 Transforming growth factor beta-2 Transforming growth factor beta-2 proprotein transforming growth factor beta 2; BSC-1 cell growth inhibitor; Cetermin; G-TSF; Glioblastoma-derived T-cell suppressor factor; LAP; Latency-associated peptide; polyergin; prepro-transforming growth factor beta-2; TGF-beta-2; Transforming growth factor beta-2; Transforming growth factor beta-2 proprotein; transforming growth factor, beta 2
Gene Aliases: BB105277; LDS4; TGF-beta2; Tgfb-2; TGFB2
UniProt ID: (Human) P61812, (Mouse) P27090
Entrez Gene ID: (Human) 7042, (Mouse) 21808
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.