Invitrogen
Manufacturer Code:MA516949
Catalog # PIMA516949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TB21 |
Immunogen | Human Transforming Growth Factor Beta 1 from platelets |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C or -20°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CED DPD1 LAP TGFB TGFbetaTGF-beta 1 protein TGF-beta-1 latency-associated peptide transforming growth factor beta-1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF-beta 1; TGF-beta-1; Transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1
Gene Aliases: CED; DPD1; LAP; TGF-beta1; TGFB; Tgfb-1; TGFB1; TGFbeta; TGFbeta1
UniProt ID: (Human) P01137, (Sheep) P50414, (Rat) P17246, (Mouse) P04202
Entrez Gene ID: (Human) 7040, (Rabbit) 100008645, (Sheep) 443417, (Rat) 59086, (Mouse) 21803
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.