Invitrogen
Manufacturer Code:MA515065
Catalog # PIMA515065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | F.888.7 |
Immunogen | Synthetic peptide corresponding to a region in the carboxy terminus of TGF-ß1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: cytokine; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF-beta 1; TGF-beta-1; Transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; unnamed protein product
Gene Aliases: CED; DPD1; LAP; TGF-BETA-1; TGF-beta1; TGFB; Tgfb-1; TGFB1; TGFbeta; TGFbeta1
UniProt ID: (Human) P01137, (Pig) P07200, (Rat) P17246, (Mouse) P04202
Entrez Gene ID: (Human) 7040, (Pig) 397078, (Rat) 59086, (Mouse) 21803
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.