Invitrogen
Manufacturer Code:MA121479
Catalog # PIMA121479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2E6 |
Immunogen | TGF-beta 1 from human platelets |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CED DPD1 LAP TGFB TGFbeta; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; TGF-beta 1; TGF-beta-1; Transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1 (Camurati-Engelmann disease)
Gene Aliases: CED; DPD1; LAP; TGFB; TGFB1; TGFbeta
UniProt ID: (Human) P01137, (Sheep) P50414
Entrez Gene ID: (Human) 7040, (Sheep) 443417
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.