Invitrogen
Manufacturer Code:PA1197
Catalog # PIPA1197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human TGF beta-1 protein (aa 279-390) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C |
TGF beta-1 Antibody, PA1-197, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.
Protein Aliases: LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; TGF-beta-1; TGFB TGFb-1 DPD1 Transforming Growth Factor Transforming Growth Factor Beta-1 TGF-beta-1 CED; Transforming growth factor beta-1; Transforming growth factor beta-1 proprotein
Gene Aliases: CED; DPD1; LAP; TGFB; TGFB1; TGFbeta
UniProt ID: (Human) P01137
Entrez Gene ID: (Human) 7040
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.